Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd99l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134459.1 | NP_604454.1 | 1, 2 | 138 | ||
XM_006251676.2 | XP_006251738.1 | 1, 2 | 174 | |||
XM_008770596.1 | XP_008768818.1 | 1, 2 | 164 | |||
GenBank mRNA | AB031014.1 | - | 1, 2 | 138 | ||
AF481858.1 | - | 1, 2 | 85 |
Gene Symbol: RGD1559441 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237887.2 | XP_006237949.1 | 1, 2 | 154 |