Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osgin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138504.2 | NP_612513.1 | 5, 6 | 747 | ||
XM_006255706.2 | XP_006255768.1 | 8, 9 | 806 | |||
XM_006255707.2 | XP_006255769.1 | 4, 5 | 842 | |||
XM_008772594.1 | XP_008770816.1 | 3, 4 | 668 | |||
GenBank mRNA | AF549441.1 | - | 5, 6 | 747 | ||
AF549442.1 | - | 4, 5 | 648 | |||
AY081218.1 | - | 3, 4 | 396 | |||
BC091111.1 | - | 5, 6 | 743 | 7106386 |