Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016248.3 | NP_057332.1 | 4 | 312 | ||
XM_005266247.1 | XP_005266304.1 | 4 | 313 | |||
XM_005266248.3 | XP_005266305.1 | 4 | 341 | |||
XM_005266249.3 | XP_005266306.1 | 4 | 313 | |||
XM_005266250.3 | XP_005266307.1 | 4 | 313 | |||
XM_011534903.2 | XP_011533205.1 | 4 | 309 | |||
XM_011534904.1 | XP_011533206.1 | 4 | 505 | |||
XM_011534905.1 | XP_011533207.1 | 3 | 263 | |||
XM_011534906.2 | XP_011533208.1 | 3 | 481 | |||
XM_017020381.1 | XP_016875870.1 | 4 | 363 | |||
XM_017020382.1 | XP_016875871.1 | 3 | 288 | |||
XM_017020383.1 | XP_016875872.1 | 3 | 476 | |||
GenBank mRNA | AF176555.1 | - | 4 | 312 | ||
CR627087.1 | - | 3 | 475 |