Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ero1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138528.1 | NP_612537.1 | 1, 2 | 135 | ||
GenBank mRNA | AF489855.1 | - | 1, 2 | 122 | ||
AY071924.1 | - | 1, 2 | 135 |