Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acmsd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134372.1 | NP_599199.1 | 207 | |||
GenBank mRNA | AB069781.1 | - | 207 |