Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr7a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134407.1 | NP_599234.1 | 4 | 662 | ||
GenBank mRNA | AB037424.1 | - | 4 | 662 | ||
AF503514.1 | - | 4 | 700 | |||
AJ271883.1 | - | 4 | 665 | |||
BC061816.1 | - | 4 | 665 | 5598724 | ||
FQ230711.1 | - | 1 | 71 | |||
FQ231883.1 | - | 1 | 71 |