Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd99l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134459.1 | NP_604454.1 | 2 | 170 | ||
GenBank mRNA | AB031014.1 | - | 2 | 170 |
Gene Symbol: RGD1559441 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237887.2 | XP_006237949.1 | 2 | 186 | ||
XM_008763576.1 | XP_008761798.1 | 7 | 954 | |||
XM_008763577.1 | XP_008761799.1 | 7 | 1234 |