Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgll | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138502.2 | NP_612511.1 | 5, 6 | 605 | ||
XM_008763053.1 | XP_008761275.1 | 5, 6 | 1263 | |||
GenBank mRNA | AY081195.1 | - | 4, 5 | 488 | ||
BC107920.1 | - | 5, 6 | 605 | 7446100 | ||
FQ227013.1 | - | 5, 6 | 1231 |