Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138508.4 | NP_612517.2 | 4, 5 | 353 | ||
GenBank mRNA | BC081713.1 | - | 4, 5 | 366 | 7109192 | |
M57453.1 | - | 4, 5 | 353 | |||
M58287.1 | - | 3, 4 | 239 | |||
M62763.1 | - | 4, 5 | 380 | |||
X60654.1 | - | 4, 5 | 333 |