Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gzmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138517.3 | NP_612526.2 | 4 | 494 | ||
GenBank mRNA | BC127475.1 | - | 4 | 474 | 7124652 | |
FQ220462.1 | - | 1 | 41 | |||
FQ220750.1 | - | 4 | 516 | |||
FQ221957.1 | - | 4 | 516 | |||
FQ228927.1 | - | 4 | 516 | |||
X66693.1 | - | 496 |
Gene Symbol: Gzmbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773505.1 | XP_008771727.1 | 4 | 489 |
Gene Symbol: Gzmbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728257.4 | XP_002728303.2 | 4 | 562 |
Gene Symbol: LOC102553861 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770729.1 | XP_008768951.1 | 2 | 141 |