Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsbg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134389.1 | NP_599216.1 | 6 | 717 | ||
XM_008766203.1 | XP_008764425.1 | 7 | 993 | |||
XM_008766204.1 | XP_008764426.1 | 7 | 4702 | |||
GenBank mRNA | AF208125.1 | - | 6 | 717 |