Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pacs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134406.1 | NP_599233.1 | 5 | 886 | ||
XM_006230657.2 | XP_006230719.1 | 5 | 975 | |||
GenBank mRNA | AF076183.1 | - | 5 | 886 | ||
AF076184.1 | - | 5 | 886 |