Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138514.1 | NP_612523.1 | 4 | 628 | ||
GenBank mRNA | BC088105.1 | - | 4 | 650 | 7372890 | |
BC098903.1 | - | 4 | 629 | 7373446 | ||
FQ218915.1 | - | 2 | 297 | |||
J02861.1 | - | 4 | 630 | |||
M32277.1 | - | 4 | 628 | |||
M33994.1 | - | 4 | 617 |