Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138831.1 | NP_620186.1 | 1 | 511 | ||
XM_008772974.1 | XP_008771196.1 | 1 | 550 | |||
XM_008772975.1 | XP_008771197.1 | 1 | 549 | |||
GenBank mRNA | AB047324.1 | - | 1 | 511 |