Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sftpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138842.1 | NP_620197.1 | 2 | 111 | ||
XM_006236657.2 | XP_006236719.1 | 2 | 146 | |||
GenBank mRNA | BC072466.1 | - | 2 | 110 | 7100971 | |
X14778.1 | - | 2 | 111 |