Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpm6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138846.1 | NP_620201.1 | 3 | 575 | ||
XM_006256849.2 | XP_006256911.1 | 4 | 1114 | |||
XM_006256850.2 | XP_006256912.1 | 4 | 1116 | |||
XM_006256851.2 | XP_006256913.1 | 3 | 638 | |||
XM_008773151.1 | XP_008771373.1 | 3 | 627 | |||
GenBank mRNA | AB036421.1 | - | 3 | 575 |