Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spdya | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138855.1 | NP_620210.1 | 2 | 273 | ||
XM_006239766.1 | XP_006239828.1 | 2 | 507 | |||
XM_006239767.2 | XP_006239829.1 | 2 | 471 | |||
XM_006239768.2 | XP_006239830.1 | 2 | 338 | |||
XM_008764502.1 | XP_008762724.1 | 2 | 1173 | |||
GenBank mRNA | AF492385.1 | - | 2 | 273 | ||
BC078729.1 | - | 1 | 196 | 7134092 |