Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptgr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138863.2 | NP_620218.1 | 6 | 463 | ||
GenBank mRNA | BC089775.1 | - | 6 | 463 | 7307365 | |
FQ218841.1 | - | 6 | 482 | |||
FQ220898.1 | - | 6 | 484 | |||
FQ221589.1 | - | 6 | 479 | |||
FQ222466.1 | - | 6 | 481 | |||
FQ229147.1 | - | 6 | 478 | |||
FQ229679.1 | - | 6 | 479 | |||
U66322.2 | - | 5 | 428 |