Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golgb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138885.1 | NP_620240.1 | 4 | 404 | ||
XM_006248379.2 | XP_006248441.1 | 4 | 2274 | |||
XM_006248380.2 | XP_006248442.1 | 3 | 2122 | |||
XM_008768698.1 | XP_008766920.1 | 3 | 1467 | |||
XM_008768699.1 | XP_008766921.1 | 3 | 1467 | |||
XM_008768700.1 | XP_008766922.1 | 3 | 1467 | |||
XM_008768701.1 | XP_008766923.1 | 3 | 1467 | |||
XM_008768702.1 | XP_008766924.1 | 2 | 116 | |||
XM_008768703.1 | XP_008766925.1 | 3 | 1468 | |||
GenBank mRNA | D25543.1 | - | 4 | 404 | ||
FQ230890.1 | - | 4 | 405 | |||
FQ230926.1 | - | 4 | 382 |