Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnrc6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138845.2 | NP_620200.2 | 25 | 5760 | ||
XM_006242061.2 | XP_006242123.1 | 22 | 6040 | |||
XM_006242063.2 | XP_006242125.1 | 21 | 5868 | |||
XM_006242064.2 | XP_006242126.1 | 20 | 5709 | |||
GenBank mRNA | AF275151.1 | - | 17 | 2515 | ||
BC061751.1 | - | 9 | 1436 | |||
FQ226824.1 | - | 2 | 630 |