Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2b5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138866.2 | NP_620221.2 | 2 | 296 | ||
GenBank mRNA | BC085698.1 | - | 2 | 296 | 7193458 | |
U19516.1 | - | 2 | 294 |