Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128160.1 | NP_001121632.1 | 15 | 3021 | ||
NM_001128161.1 | NP_001121633.1 | 17 | 2748 | |||
NM_014517.4 | NP_055332.3 | 16 | 3129 | |||
GenBank mRNA | AF198487.1 | - | 16 | 2774 | ||
AK023274.1 | - | 15 | 2650 | |||
AK023278.1 | - | 10 | 1931 | |||
AK023322.1 | - | 1 | 880 | |||
AK027177.1 | - | 1 | 1705 | |||
AL133032.1 | - | 1 | 815 | |||
AL831847.1 | - | 1 | 815 | |||
BC047235.1 | - | 17 | 3128 | 5262788 | ||
CR749798.1 | - | 2 | 6378 |