Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hyou1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034028.1 | NP_001029200.1 | 3, 4 | 244 | ||
NM_138867.2 | NP_620222.2 | 3, 4 | 396 | |||
XM_006242884.2 | XP_006242946.1 | 3, 4 | 341 | |||
XM_006242885.2 | XP_006242947.1 | 3, 4 | 277 | |||
XM_008766143.1 | XP_008764365.1 | 3, 4 | 323 | |||
GenBank mRNA | BC065310.1 | - | 3, 4 | 287 | 5621638 | |
CB691248.1 | - | 3, 4 | 252 | |||
U41853.1 | - | 3, 4 | 396 |