Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138888.1 | NP_620243.1 | 3, 4 | 248 | ||
GenBank mRNA | AF495522.1 | - | 3, 4 | 248 | ||
AY321339.1 | - | 264 | ||||
BC168144.1 | - | 3, 4 | 266 | 8372591 | ||
FQ223455.1 | - | 1, 2 | 22 |