Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033867.1 | NP_001029039.1 | 1 | 475 | ||
NM_138916.2 | NP_620271.1 | 1 | 475 | |||
XM_006247250.2 | XP_006247312.1 | 1 | 1029 | |||
XM_006247251.2 | XP_006247313.1 | 1 | 1029 | |||
XM_006247253.2 | XP_006247315.1 | 1 | 1028 | |||
GenBank mRNA | AY072294.1 | - | 1 | 475 | ||
AY962568.1 | - | 1 | 475 |