Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138921.1 | NP_620276.1 | 10 | 1070 | ||
GenBank mRNA | AF335571.1 | - | 10 | 1070 | ||
BC062038.1 | - | 7 | 612 | 5598497 |