Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138538.1 | NP_612547.1 | 15, 16 | 1827 | ||
XM_006250141.2 | XP_006250203.1 | 14, 15 | 1837 | |||
XM_006250142.2 | XP_006250204.1 | 14, 15 | 1837 | |||
GenBank mRNA | AF201839.1 | - | 15, 16 | 1827 | ||
D14076.1 | - | 14, 15 | 1797 |