Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZKSCAN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318082.1 | NP_001305011.1 | 5 | 944 | ||
NM_014569.3 | NP_055384.1 | 5 | 1003 | |||
NM_145102.3 | NP_659570.1 | 5 | 882 | |||
XM_011515999.2 | XP_011514301.1 | 4 | 543 | |||
XM_017011918.1 | XP_016867407.1 | 7 | 1341 | |||
XM_017011919.1 | XP_016867408.1 | 6 | 1384 | |||
XM_017011920.1 | XP_016867409.1 | 4 | 1546 | |||
XM_017011921.1 | XP_016867410.1 | 5 | 1312 | |||
XM_017011922.1 | XP_016867411.1 | 5 | 971 | |||
XM_017011925.1 | XP_016867414.1 | 4 | 488 | |||
GenBank mRNA | AB023232.1 | - | 5 | 944 | ||
AB742318.1 | - | 4 | 433 | |||
AF170025.1 | - | 5 | 856 | |||
AK027714.1 | - | 5 | 972 | |||
AK096552.1 | - | 5 | 1541 | |||
BC030790.2 | - | 5 | 913 | 4827870 |