Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138911.3 | NP_620266.1 | 3 | 365 | ||
GenBank mRNA | BC061529.1 | - | 3 | 366 | 6887497 | |
Y15068.1 | - | 2 | 300 |