Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138912.2 | NP_620267.2 | 2 | 912 | ||
XM_006253215.2 | XP_006253277.1 | 2 | 1478 | |||
XM_006253216.2 | XP_006253278.1 | 2 | 799 | |||
GenBank mRNA | BC072514.1 | - | 2 | 912 | 7104413 | |
Y18208.1 | - | 2 | 775 |