Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033080.1 | NP_001028252.1 | 2 | 657 | ||
NM_014626.3 | NP_055441.2 | 1 | 545 | |||
GenBank mRNA | AY702304.1 | - | 1 | 522 | ||
AY703480.1 | - | 2 | 657 | |||
BC067461.1 | - | 1 | 522 | |||
BC067462.1 | - | 1 | 522 | |||
BC067463.1 | - | 1 | 522 | |||
DN693314.1 | - | 1 | 545 |
Set Membership: |
G-protein coupled receptor |