Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prokr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138978.1 | NP_620434.1 | 1 | 245 | ||
XM_006235059.2 | XP_006235121.1 | 3 | 651 | |||
GenBank mRNA | AY089975.1 | - | 1 | 245 |
Set Membership: |
G-protein coupled receptor |