Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Muc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768788.1 | XP_008767010.1 | 2, 3 | 687 | ||
GenBank mRNA | U89744.1 | - | 1, 2 | 649 |