Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xrcc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139080.2 | NP_620780.2 | 3 | 186 | ||
XM_008765671.1 | XP_008763893.1 | 3 | 179 | |||
GenBank mRNA | AB066102.1 | - | 2 | 170 | ||
BC078718.1 | - | 3 | 186 | 7115290 |