Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139108.2 | NP_620808.1 | 257 | |||
XM_006241924.2 | XP_006241986.1 | 2 | 570 | |||
XM_006241925.2 | XP_006241987.1 | 2 | 560 | |||
XM_006241926.2 | XP_006241988.1 | 2 | 548 | |||
XM_008765544.1 | XP_008763766.1 | 2 | 591 | |||
GenBank mRNA | AF290194.1 | - | 262 |
Gene Symbol: LOC100910472 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241915.2 | XP_006241977.1 | 2 | 570 | ||
XM_006241916.2 | XP_006241978.1 | 2 | 586 | |||
XM_006241917.2 | XP_006241979.1 | 2 | 560 | |||
XM_006241918.2 | XP_006241980.1 | 2 | 548 | |||
XM_008765634.1 | XP_008763856.1 | 2 | 591 |