Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC681458 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760487.1 | XP_008758709.1 | 1 | 272 |
Gene Symbol: Scd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139192.2 | NP_631931.2 | 2 | 344 | ||
XM_006231433.2 | XP_006231495.1 | 3 | 479 | |||
GenBank mRNA | AF509568.1 | - | 2 | 344 | ||
AF509569.1 | - | 2 | 344 | |||
J02585.1 | - | 2 | 398 |