Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139254.2 | NP_640347.2 | 2 | 123 | ||
GenBank mRNA | BC097281.1 | - | 2 | 103 | 7461390 | |
CF976541.1 | - | 2 | 127 |