Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139256.1 | NP_640349.1 | 2 | 315 | ||
GenBank mRNA | BC086989.1 | - | 2 | 196 | 7193864 | |
M57547.1 | - | 2 | 315 |