Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139327.1 | NP_647543.1 | 2 | 194 | ||
XM_006256156.1 | XP_006256218.1 | 3 | 341 | |||
XM_006256157.2 | XP_006256219.1 | 4 | 454 | |||
XM_006256158.2 | XP_006256220.1 | 3 | 383 | |||
XM_006256161.1 | XP_006256223.1 | 3 | 308 | |||
XM_006256162.2 | XP_006256224.1 | 3 | 344 | |||
XM_006256163.2 | XP_006256225.1 | 3 | 719 | |||
XM_006256164.1 | XP_006256226.1 | 3 | 350 | |||
GenBank mRNA | AF507966.1 | - | 2 | 194 | ||
AF511040.1 | - | 2 | 181 |