Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139097.3 | NP_620797.1 | 3 | 586 | ||
XM_006243102.1 | XP_006243164.1 | 3 | 668 | |||
XM_006243103.1 | XP_006243165.1 | 3 | 668 | |||
XM_008766114.1 | XP_008764336.1 | 3 | 668 | |||
XM_008766115.1 | XP_008764337.1 | 2 | 235 | |||
GenBank mRNA | AF378093.1 | - | 3 | 285 | ||
AJ243395.2 | - | 3 | 571 | |||
BC070899.1 | - | 3 | 586 | 7100476 |