Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il11ra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139116.1 | NP_620816.1 | 9 | 895 | ||
XM_006238021.2 | XP_006238083.1 | 8 | 1613 | |||
XM_006238022.2 | XP_006238084.1 | 8 | 1033 | |||
XM_006238023.2 | XP_006238085.1 | 9 | 1515 | |||
GenBank mRNA | AF347936.1 | - | 9 | 895 | ||
BC070921.1 | - | 9 | 890 | 7105297 |