Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139261.1 | NP_640354.1 | 5 | 866 | ||
XM_006228248.1 | XP_006228310.1 | 5 | 1093 | |||
XM_006228249.2 | XP_006228311.1 | 5 | 858 | |||
XM_006228250.2 | XP_006228312.1 | 5 | 922 | |||
GenBank mRNA | AF187860.1 | - | 5 | 866 | ||
BC072541.1 | - | 5 | 902 | 7103744 | ||
FQ232199.1 | - | 5 | 922 |