Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270788.1 | NP_001257717.1 | 5, 6 | 729 | ||
NM_139100.2 | NP_620800.2 | 5, 6 | 726 | |||
GenBank mRNA | BC070918.1 | - | 5, 6 | 668 | 7104964 | |
FQ210190.1 | - | 5, 6 | 680 | |||
FQ213750.1 | - | 5, 6 | 685 | |||
FQ214698.1 | - | 5, 6 | 683 | |||
FQ215315.1 | - | 5, 6 | 683 | |||
FQ216349.1 | - | 5, 6 | 682 | |||
FQ216421.1 | - | 5, 6 | 683 | |||
FQ216474.1 | - | 5, 6 | 691 | |||
FQ217707.1 | - | 5, 6 | 683 | |||
FQ220626.1 | - | 5, 6 | 667 | |||
FQ223916.1 | - | 5, 6 | 683 | |||
FQ227446.1 | - | 5, 6 | 680 | |||
FQ229113.1 | - | 5, 6 | 680 | |||
FQ230627.1 | - | 5, 6 | 682 | |||
FQ232293.1 | - | 3, 4 | 485 | |||
FQ232835.1 | - | 5, 6 | 676 | |||
FQ234854.1 | - | 5, 6 | 680 | |||
M23984.1 | - | 4, 5 | 645 |