Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749863.3 | XP_003749911.1 | 4 | 898 | ||
XM_006237585.2 | XP_006237647.1 | 4 | 882 | |||
XM_006237588.2 | XP_006237650.1 | 5 | 890 | |||
XM_006237590.2 | XP_006237652.1 | 4 | 496 | |||
XM_006237591.2 | XP_006237653.1 | 4 | 676 | |||
XM_008763453.1 | XP_008761675.1 | 4 | 826 | |||
GenBank mRNA | FQ213730.1 | - | 3 | 641 |