Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sipa1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139330.1 | NP_647546.1 | 2815 | |||
GenBank mRNA | AF026504.1 | - | 2815 | |||
AY043226.1 | - | 2378 |