Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc34a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139338.2 | NP_647554.1 | 11 | 1247 | ||
XM_006233558.2 | XP_006233620.1 | 12 | 1460 | |||
XM_006233559.2 | XP_006233621.1 | 12 | 1576 | |||
XM_006233560.2 | XP_006233622.1 | 12 | 1437 | |||
XM_006233561.2 | XP_006233623.1 | 11 | 1350 | |||
XM_006233562.2 | XP_006233624.1 | 11 | 1466 | |||
XM_006233564.2 | XP_006233626.1 | 11 | 1330 | |||
XM_006233565.2 | XP_006233627.1 | 12 | 1436 | |||
XM_006233566.2 | XP_006233628.1 | 11 | 1442 | |||
XM_006233567.2 | XP_006233629.1 | 11 | 1318 | |||
XM_008761577.1 | XP_008759799.1 | 12 | 1525 | |||
GenBank mRNA | AB077042.1 | - | 11 | 1252 |