Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144740.3 | NP_653341.2 | 12 | 1682 | ||
XM_006229545.2 | XP_006229607.1 | 12 | 2442 | |||
XM_006229546.2 | XP_006229608.1 | 11 | 2379 | |||
XM_006229547.2 | XP_006229609.1 | 11 | 2340 | |||
XM_006229548.2 | XP_006229610.1 | 12 | 2442 | |||
GenBank mRNA | AF401635.1 | - | 12 | 1692 | ||
AY742900.1 | - | 1695 |