Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144748.1 | NP_653349.1 | 6 | 904 | ||
XM_008759677.1 | XP_008757899.1 | 6 | 889 | |||
XM_008759678.1 | XP_008757900.1 | 6 | 926 | |||
XM_008759679.1 | XP_008757901.1 | 6 | 927 | |||
GenBank mRNA | AF062389.1 | - | 6 | 904 | ||
BC078721.1 | - | 6 | 881 | 7110246 |