Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145774.2 | NP_665717.1 | 1, 2 | 256 | ||
GenBank mRNA | BC070513.1 | - | 1, 2 | 266 | 7099841 | |
M94043.1 | - | 1, 2 | 259 |