Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113422.1 | NP_001106893.1 | 5 | 1079 | ||
NM_145775.2 | NP_665718.2 | 5 | 1049 | |||
GenBank mRNA | BC062047.1 | - | 5 | 1079 | 5598020 | |
DN948099.1 | - | 1 | 155 | |||
M25804.1 | - | 5 | 1049 |